2-Dimethylaminoethanethiol hydrochloride, 95%, Thermo Scientific Chemicals
2-Dimethylaminoethanethiol hydrochloride, 95%, Thermo Scientific Chemicals
2-Dimethylaminoethanethiol hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Dimethylaminoethanethiol hydrochloride, 95%, Thermo Scientific Chemicals

Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 115811000
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS13242-44-9
IUPAC Namehydrogen 2-(dimethylamino)ethane-1-thiol chloride
Molecular FormulaC4H12ClNS
InChI KeyNRVFDGZJTPCULU-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)White to off-white
Titration Iodimetric>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only