9,10-Dimethylanthracene, 97%, Thermo Scientific Chemicals
9,10-Dimethylanthracene, 97%, Thermo Scientific Chemicals
9,10-Dimethylanthracene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

9,10-Dimethylanthracene, 97%, Thermo Scientific Chemicals

CAS: 781-43-1 | Molecular Formula: C16H14 | Molecular Weight: 206.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1159700101 gGlass bottle
Catalog number 115970010
Price (USD)
118.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
118.00
Each
Chemical Identifiers
CAS781-43-1
IUPAC Name9,10-dimethylanthracene
Molecular FormulaC16H14
InChI KeyJTGMTYWYUZDRBK-UHFFFAOYSA-N
SMILESCC1=C2C=CC=CC2=C(C)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point180°C to 185°C
GC>=96.0 %
Appearance (Form)Crystalline powder or crystals
Appearance (Color)Yellow

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only