N,N-Dimethylethanolamine, 99%
N,N-Dimethylethanolamine, 99%
N,N-Dimethylethanolamine, 99%
Thermo Scientific Chemicals

N,N-Dimethylethanolamine, 99%

CAS: 108-01-0 | C4H11NO | 89.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
25 L
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 116180010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-01-0
IUPAC Name2-(dimethylamino)ethan-1-ol
Molecular FormulaC4H11NO
InChI KeyUEEJHVSXFDXPFK-UHFFFAOYSA-N
SMILESCN(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Water=<0.1 %
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only