unsym-Dimethylethylenediamine, 99%
unsym-Dimethylethylenediamine, 99%
unsym-Dimethylethylenediamine, 99%
Thermo Scientific Chemicals

unsym-Dimethylethylenediamine, 99%

CAS: 108-00-9 | C4H12N2 | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 116205000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-00-9
IUPAC Name(2-aminoethyl)dimethylamine
Molecular FormulaC4H12N2
InChI KeyDILRJUIACXKSQE-UHFFFAOYSA-N
SMILESCN(C)CCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Water=<1 %
Appearance (Form)Liquid
Refractive index1.4255 to 1.4275 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only