N,N-Dimethyl-3-nitroaniline, 98%
N,N-Dimethyl-3-nitroaniline, 98%
N,N-Dimethyl-3-nitroaniline, 98%
Thermo Scientific Chemicals

N,N-Dimethyl-3-nitroaniline, 98%

CAS: 619-31-8 | C8H10N2O2 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 116470050
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.80
Save 5.15
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.80
Save 5.15
Each
Chemical Identifiers
CAS619-31-8
IUPAC NameN,N-dimethyl-3-nitroaniline
Molecular FormulaC8H10N2O2
InChI KeyCJDICMLSLYHRPT-UHFFFAOYSA-N
SMILESCN(C)C1=CC=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point57°C to 61°C
Appearance (Color)Orange to orange-red
Titration with HClO4>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only