2,5-Dimethylphenol, 99+%
2,5-Dimethylphenol, 99+%
2,5-Dimethylphenol, 99+%
Thermo Scientific Chemicals

2,5-Dimethylphenol, 99+%

CAS: 95-87-4 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
Packaging:
Glass bottle
Catalog number 116545000
Price (USD)
239.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
239.00
Each
Chemical Identifiers
CAS95-87-4
IUPAC Name2,5-dimethylphenol
Molecular FormulaC8H10O
InChI KeyNKTOLZVEWDHZMU-UHFFFAOYSA-N
SMILESCC1=CC=C(C)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Infrared spectrumConforms
GC>=99.0 %
Sulfated ash=<0.2 %
Melting point>=75°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only