3,5-Dimethylpyrazole, 99%
3,5-Dimethylpyrazole, 99%
3,5-Dimethylpyrazole, 99%
Thermo Scientific Chemicals

3,5-Dimethylpyrazole, 99%

CAS: 67-51-6 | C5H8N2 | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 116701000
Price (USD)
68.65
Online exclusive
75.90
Save 7.25
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
68.65
Online exclusive
75.90
Save 7.25
Each
Chemical Identifiers
CAS67-51-6
IUPAC Name3,5-dimethyl-1H-pyrazole
Molecular FormulaC5H8N2
InChI KeySDXAWLJRERMRKF-UHFFFAOYSA-N
SMILESCC1=CC(C)=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point106°C to 109°C
Appearance (Color)White to off-white
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only