1,4-Dioxane, 99+%, extra pure, stabilized
1,4-Dioxane, 99+%, extra pure, stabilized
1,4-Dioxane, 99+%, extra pure, stabilized
Thermo Scientific Chemicals

1,4-Dioxane, 99+%, extra pure, stabilized

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Metal Drum
Catalog number 117110250
Price (EUR)
1 224,00
Each
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (EUR)
1 224,00
Each
Chemical Identifiers
CAS123-91-1
IUPAC Name1,4-dioxane
Molecular FormulaC4H8O2
InChI KeyRYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILESC1COCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Peroxides=<0.005 % (as H2O2)
Color scale=<20 APHA
Residue after evaporation=<0.01 %
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only