N,N'-Difenilbencidina, 97 %, Thermo Scientific Chemicals
N,N'-Difenilbencidina, 97 %, Thermo Scientific Chemicals
N,N'-Difenilbencidina, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N'-Difenilbencidina, 97 %, Thermo Scientific Chemicals

CAS: 531-91-9 | C24H20N2 | 336.44 g/mol
Have Questions?
Número de catálogoCantidadEnvase
1172100505 gFrasco de vidrio
Número de catálogo 117210050
Precio (USD)
-
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS531-91-9
IUPAC NameN4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Molecular FormulaC24H20N2
InChI KeyFDRNXKXKFNHNCA-UHFFFAOYSA-N
SMILESN(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Crystalline powder
Melting point246°C to 250°C
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Color)Grey to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicador
RUO – Research Use Only