trans,trans-1,4-Diphenyl-1,3-butadiene, 99%
trans,trans-1,4-Diphenyl-1,3-butadiene, 99%
trans,trans-1,4-Diphenyl-1,3-butadiene, 99%
Thermo Scientific Chemicals

trans,trans-1,4-Diphenyl-1,3-butadiene, 99%

CAS: 538-81-8 | C16H14 | 206.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 117220250
Price (USD)
92.65
Special offer
Online exclusive
Ends: 28-Dec-2026
109.00
Save 16.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
Ends: 28-Dec-2026
109.00
Save 16.35
Each
Chemical Identifiers
CAS538-81-8
IUPAC Name[(1E,3E)-4-phenylbuta-1,3-dien-1-yl]benzene
Molecular FormulaC16H14
InChI KeyJFLKFZNIIQFQBS-FNCQTZNRSA-N
SMILESC(\C=C\C1=CC=CC=C1)=C/C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to light yellow
Melting point151°C to 154°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only