CellsDirect™ Two-Step qRT-PCR Kit - FAQs
View additional product information for CellsDirect™ Two-Step qRT-PCR Kit - FAQs (11737030)
1 product FAQs found
I want to perform qRT-PCR using cDNA that I synthesize directly from cells. Can I use the SuperScript III CellsDirect cDNA Kit for end-point PCR and the Platinum Quantitative PCR SuperMix-UDG kit?
We would recommend using our Cells-to-CT kits for best qRT-PCR results. The CellsDirect kits for end-point PCR have not been optimized for qRT-PCR.