Search
Search

| 카탈로그 번호 | 반응 수 |
|---|---|
| 11750350 | 500 Reactions |
| 11750150 | 50 Reactions |
The SuperScript IV CellsDirect cDNA Synthesis Kit is optimized for the synthesis of first-strand cDNA directly from a mammalian cell lysate without first isolating RNA. Cell lysis and reverse transcription are performed in the same tube, and the resulting first-strand cDNA is ready to use in PCR and qPCR.
Features of the SuperScript IV CellsDirect Synthesis Kit
Features of the SuperScript IV Reverse Transcriptase included in this kit
SuperScript IV reverse transcriptase is a top choice for all RT-PCR and qRT-PCR applications. It is a proprietary MMLV mutant with superior robustness and reliability in RT reactions. It is significantly improved over SuperScript III reverse transcriptase in inhibitor resistance, processivity, and reaction speed, while retaining all the benefits of the previous enzyme, including increased thermostability, highly efficient full-length cDNA synthesis, and reduced RNase activity.
How it works
In traditional RT-PCR, RNA is first isolated from cells in a time-consuming procedure that can lead to a loss of material. Using the SuperScript IV CellsDirect cDNA Synthesis Kit, the cells are lysed and the cDNA is generated from the lysate in a single tube with minimal handling and no sample loss. DNase I treatment is an optional step and can be used to eliminate genomic DNA prior to first-strand synthesis. This kit has been optimized for small cell samples, ranging from 10,000 cells down to a single cell.
• SuperScript IV CellsDirect Lysis Solution (3 x 9 mL)
• SuperScript IV CellsDirect Stop Solution (2 x 1.25 mL)
• Lysis Enhancer (1 x 250 μL)
• DNase I (1 x 500 μL)
• SuperScript IV RT Master Mix (4 x 1 mL)
• SuperScript IV No RT Control (4 x 1 mL)
Store at –20°C.