5,5'-Dithiobis-(2-nitrobenzoic acid), 99%
5,5'-Dithiobis-(2-nitrobenzoic acid), 99%
5,5'-Dithiobis-(2-nitrobenzoic acid), 99%
Thermo Scientific Chemicals

5,5'-Dithiobis-(2-nitrobenzoic acid), 99%

CAS: 69-78-3 | C14H8N2O8S2 | 396.34 g/mol
数量:
5 g
25 g
パッケージ:
Glass Bottle
製品番号(カタログ番号) 117540250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS69-78-3
Appearance (Form)Crystalline powder
Loss on drying=<0.5 %
Appearance (Color)Off-white to yellow
Infrared spectrumConforms
Melting point240°C to 245°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only