Dodecylamine, 98%
Dodecylamine, 98%
Dodecylamine, 98%
Thermo Scientific Chemicals

Dodecylamine, 98%

CAS: 124-22-1 | C12H27N | 185.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number 117660010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS124-22-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Low melting crystalline solid
Melting point27°C to 31°C
GC>=97.5 % (Alkyl chain distribution C12)
Titration with HClO4>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only