Epichlorohydrin, 99%
Epichlorohydrin, 99%
Epichlorohydrin, 99%
Thermo Scientific Chemicals

Epichlorohydrin, 99%

CAS: 106-89-8 | C3H5ClO | 92.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 117780025
Price (USD)
129.65
Special offer
Online exclusive
Ends: 28-Dec-2026
152.00
Save 22.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
129.65
Special offer
Online exclusive
Ends: 28-Dec-2026
152.00
Save 22.35
Each
Chemical Identifiers
CAS106-89-8
IUPAC Name2-(chloromethyl)oxirane
Molecular FormulaC3H5ClO
InChI KeyBRLQWZUYTZBJKN-UHFFFAOYNA-N
SMILESClCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.02 % (K.F.)
Color scale=<15 APHA
Infrared spectrumConforms
Appearance (Form)Clear liquid
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only