Library Efficiency® DB3.1™ Competent Cells -"DISCONTINUED" - FAQs
Library Efficiency® DB3.1™ Competent Cells -"DISCONTINUED" - FAQs
View additional product information for Library Efficiency® DB3.1™ Competent Cells -"DISCONTINUED" - FAQs (11782018)
1 product FAQs found
I just found out that Library Efficiency DB3.1 and One Shot ccdB Survival T1R cells have been discontinued. Do you offer an alternative for propagating ccdB-containing plasmids?
We offer One Shot ccdB Survival 2 T1R cells (Cat. No. A10460) for propagating ccdB-containing plasmids.