Ethoxymethylenemalononitrile, 98%
Ethoxymethylenemalononitrile, 98%
Ethoxymethylenemalononitrile, 98%
Thermo Scientific Chemicals

Ethoxymethylenemalononitrile, 98%

CAS: 123-06-8 | C6H6N2O | 122.127 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Catalog number 117921000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS123-06-8
IUPAC Name2-(ethoxymethylidene)propanedinitrile
Molecular FormulaC6H6N2O
InChI KeyOEICGMPRFOJHKO-UHFFFAOYSA-N
SMILESCCOC=C(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Yellow to orange-brown
Melting point63°C to 68°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only