Ethyl acrylate, 99.5%, stabilized
Ethyl acrylate, 99.5%, stabilized
Ethyl acrylate, 99.5%, stabilized
Thermo Scientific Chemicals

Ethyl acrylate, 99.5%, stabilized

CAS: 140-88-5 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 117980025
Price (USD)
147.65
Special offer
Online exclusive
Ends: 28-Dec-2026
173.00
Save 25.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
147.65
Special offer
Online exclusive
Ends: 28-Dec-2026
173.00
Save 25.35
Each
Chemical Identifiers
CAS140-88-5
IUPAC Nameethyl prop-2-enoate
Molecular FormulaC5H8O2
InChI KeyJIGUQPWFLRLWPJ-UHFFFAOYSA-N
SMILESCCOC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Stabilizer10 to 20 ppm MEHQ
GC>=99.40 %
Refractive index1.4050 to 1.4070 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only