Ethyl 3-aminocrotonate, 98.5%
Ethyl 3-aminocrotonate, 98.5%
Ethyl 3-aminocrotonate, 98.5%
Thermo Scientific Chemicals

Ethyl 3-aminocrotonate, 98.5%

CAS: 7318-00-5 | C6H11NO2 | 129.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 118035000
Price (USD)
181.65
Online exclusive
202.00
Save 20.35
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
181.65
Online exclusive
202.00
Save 20.35
Each
Chemical Identifiers
CAS7318-00-5
IUPAC Nameethyl (2E)-3-aminobut-2-enoate
Molecular FormulaC6H11NO2
InChI KeyYPMPTULBFPFSEQ-SNAWJCMRSA-N
SMILESCCOC(=O)\C=C(/C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow to orange
Appearance (Form)Low melting solid or liquid
Infrared spectrumConforms
Titration with HClO498.0 to 102.0 %
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only