Ethyl oxalyl chloride, 98%
Ethyl oxalyl chloride, 98%
Ethyl oxalyl chloride, 98%
Thermo Scientific Chemicals

Ethyl oxalyl chloride, 98%

CAS: 4755-77-5 | C4H5ClO3 | 136.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 118780250
Price (USD)
29.65
Special offer
Online exclusive
Ends: 28-Dec-2026
34.60
Save 4.95
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
29.65
Special offer
Online exclusive
Ends: 28-Dec-2026
34.60
Save 4.95
Each
Chemical Identifiers
CAS4755-77-5
IUPAC Nameethyl oxalochloridate
Molecular FormulaC4H5ClO3
InChI KeyOWZFULPEVHKEKS-UHFFFAOYSA-N
SMILESCCOC(=O)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4160 to 1.4180 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only