DMEM, low glucose, pyruvate, no glutamine, no phenol red, 500 mL - FAQs
View additional product information for DMEM, low glucose, pyruvate, no glutamine, no phenol red - FAQs (11880028)
Error:This product does not have any FAQs available.