DMEM, low glucose, pyruvate, no glutamine, no phenol red, 10 x 500 mL - FAQs
DMEM, low glucose, pyruvate, no glutamine, no phenol red, 10 x 500 mL - FAQs
View additional product information for DMEM, low glucose, pyruvate, no glutamine, no phenol red - FAQs (11054001, 11880036, 11054020)
2 product FAQs found
What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?
Manganese is not present in the formulation of our catalog DMEM media products.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do you offer media without phenol red (phenol red-free media)?
Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.