F-28/50 Fixed Angle Rotor - FAQs
View additional product information for F-28/50 Fixed Angle Rotor - FAQs (11885)
1 product FAQs found
Is it necessary to store DMEM, high glucose, pyruvate medium in the dark?
Yes, the medium should be stored in the dark because there are some components in the medium such as HEPES, Tryptophan, and Riboflavin, etc. that are sensitive to light.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.