Search
View additional product information for DMEM, low glucose, pyruvate - FAQs (11885076, 11885092, 11885084)
1 product FAQs found
Manganese is not present in the formulation of our catalog DMEM media products.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.