9-Fluorenone, 99+%
9-Fluorenone, 99+%
9-Fluorenone, 99+%
Thermo Scientific Chemicals

9-Fluorenone, 99+%

CAS: 486-25-9 | C13H8O | 180.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 119205000
Price (USD)
227.65
Special offer
Online exclusive
Ends: 28-Dec-2026
268.00
Save 40.35
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
227.65
Special offer
Online exclusive
Ends: 28-Dec-2026
268.00
Save 40.35
Each
Chemical Identifiers
CAS486-25-9
IUPAC Name9H-fluoren-9-one
Molecular FormulaC13H8O
InChI KeyYLQWCDOCJODRMT-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point81°C to 85°C
GC>=99.0 %
Appearance (Color)Yellow
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only