4-Fluorobenzenesulfonyl chloride, 98%
4-Fluorobenzenesulfonyl chloride, 98%
4-Fluorobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

4-Fluorobenzenesulfonyl chloride, 98%

CAS: 349-88-2 | C6H4ClFO2S | 194.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 119350250
Price (EUR)
50,90
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
50,90
Each
Chemical Identifiers
CAS349-88-2
IUPAC Name4-fluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H4ClFO2S
InChI KeyBFXHJFKKRGVUMU-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Titration Argentometric>=97.5 % (ex Cl)
Infrared spectrumConforms
Appearance (Form)Low melting crystalline mass
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only