DMEM, high glucose, no glutamine - FAQs
View additional product information for DMEM, high glucose, no glutamine - FAQs (11960085)
1 product FAQs found
What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?
Manganese is not present in the formulation of our catalog DMEM media products.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.