4-Fluorophenyl sulfone, 99%
4-Fluorophenyl sulfone, 99%
4-Fluorophenyl sulfone, 99%
Thermo Scientific Chemicals

4-Fluorophenyl sulfone, 99%

CAS: 383-29-9 | C12H8F2O2S | 254.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 119621000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS383-29-9
IUPAC Name1-fluoro-4-(4-fluorobenzenesulfonyl)benzene
Molecular FormulaC12H8F2O2S
InChI KeyPLVUIVUKKJTSDM-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point97°C to 100°C
Appearance (Color)White to brown
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only