Furfuryl Alcohol, 98%
Furfuryl Alcohol, 98%
Furfuryl Alcohol, 98%
Thermo Scientific Chemicals

Furfuryl Alcohol, 98%

CAS: 98-00-0 | C5H6O2 | 98.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 119790100
Price (USD)
242.65
Online exclusive
269.00
Save 26.35
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
242.65
Online exclusive
269.00
Save 26.35
Each
Chemical Identifiers
CAS98-00-0
IUPAC Name(furan-2-yl)methanol
Molecular FormulaC5H6O2
InChI KeyXPFVYQJUAUNWIW-UHFFFAOYSA-N
SMILESOCC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.3 %
Refractive index1.4850 to 1.4880 (20°C, 589 nm)
Appearance (Color)Clear yellow
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only