Gibberellic Acid, 90%
Gibberellic Acid, 90%
Gibberellic Acid, 90%
Thermo Scientific Chemicals

Gibberellic Acid, 90%

CAS: 77-06-5 | C19H22O6 | 346.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 119860010
Price (USD)
57.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.50
Save 9.85
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.50
Save 9.85
Each
Chemical Identifiers
CAS77-06-5
IUPAC Name(1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
Molecular FormulaC19H22O6
InChI KeyIXORZMNAPKEEDV-QTWFBFKQSA-N
SMILESCC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Loss on drying=<3 % (100°C, 7 hrs) (vacuum)
Arsenic (As)=<3 ppm
Infrared spectrumConforms
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only