DL-Glutamic acid monohydrate, 98+%
DL-Glutamic acid monohydrate, 98+%
DL-Glutamic acid monohydrate, 98+%
Thermo Scientific Chemicals

DL-Glutamic acid monohydrate, 98+%

CAS: 19285-83-7 | C5H9NO4·H2O | 165.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number 119931000
Price (TWD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS19285-83-7
IUPAC Name2-aminopentanedioic acid hydrate
Molecular FormulaC5H11NO5
InChI KeyOZDAOHVKBFBBMZ-UHFFFAOYNA-N
SMILESO.NC(CCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.0 %
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Powder
Water=<15.5 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only