DMEM, high glucose, pyruvate
DMEM, high glucose, pyruvate
DMEM, high glucose, pyruvate
Gibco™

DMEM, high glucose, pyruvate

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.
製品番号(カタログ番号)数量
119951151 x 5 L
119950651 x 500 mL
1199507310 x 500 mL
119950401 x 1000 mL
119950816 x 1000 mL
119951231 x 10 L
製品番号(カタログ番号) 11995115
価格(JPY)
44,200
Each
数量:
1 x 5 L
Customize this product

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:

With: High Glucose, L-glutamine, Phenol Red, Sodium Pyruvate

Without: HEPES

The complete formulation is available.

Using DMEM

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
仕様
細胞株HeLa, 293, Cos-7, and PC-12
細胞タイプMammalian Cells
濃度1X
培養環境CO2
使用対象(アプリケーション)Mammalian Cell Culture
フォーマットBag
グルコース濃度4500 mg/L
製造品質cGMP-compliant under the ISO 13485 standard
製品ラインGibco
製品タイプDMEM (Dulbecco's Modified Eagle Medium)
数量1 x 5 L
品質保持期間12 Months From Date of Manufacture
出荷条件Ambient
分類Animal Origin-free
Culture TypeMammalian Cell Culture
形状Liquid
Serum LevelStandard Serum Supplementation
無菌性Sterile-filtered
Sterilization MethodSterile-filtered
添加剤ありHigh Glucose, Glutamine, Phenol Red, Sodium Pyruvate
添加剤なしNo HEPES
Unit SizeEach
組成および保存条件
Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

よくあるご質問(FAQ)

What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?

Manganese is not present in the formulation of our catalog DMEM media products.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I see a decrease in growth of my culture. What should I do?

Try changing the medium or serum. Compare media formulations for differences in glucose, amino acids, and other components. Compare an old lot of serum with a new lot. Increase initial cell inoculums. Lastly, adapt cells sequentially to new medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

引用および参考文献 (1)

引用および参考文献
Abstract
Plaque production by the polyoma virus.
Authors:DULBECCO R, FREEMAN G,
Journal:Virology
PubMed ID:13669362
Describes the original formulation of DMEM (Dulbecco’s Modified Eagle Medium), used for plaque production by the polyoma virus.