Glutaric dialdehyde, 25% w/v solution in water
Glutaric dialdehyde, 25% w/v solution in water
Glutaric dialdehyde, 25% w/v solution in water
Thermo Scientific Chemicals

Glutaric dialdehyde, 25% w/v solution in water

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 119980010
Price (USD)
111.65
Special offer
Online exclusive
Ends: 28-Dec-2026
131.00
Save 19.35
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
111.65
Special offer
Online exclusive
Ends: 28-Dec-2026
131.00
Save 19.35
Each
Chemical Identifiers
CAS111-30-8
IUPAC Namepentanedial
Molecular FormulaC5H8O2
InChI KeySXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILESO=CCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Strength23.5 to 26.5 %
pH3.2 to 4.2
Color scale=<50 APHA
Methanol=<0.3 % (GC)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only