Odatrak Fibre-Optic to 4 to 20mA adapter allows an OdaLog L2 Logger, with fibre optic fitting, to be connected to a SCADA system - transmitting data as a 4-20mA signal.
Have Questions?
Change view
Catalog Number
Type
12-0010
Fibre Optic Cable, 25 m
12-0008
Fibre Optic Cable, 5 m
25-0356
Fibre Optic to 4-20mA Converter
3 Options
Catalog number 12-0010
Price (USD)
-
Type:
Fibre Optic Cable, 25 m
Odatrak Fibre-Optic to 4 to 20mA adapter allows an OdaLog L2 Logger, with fibre optic fitting, to be connected to a SCADA system - transmitting data as a 4-20mA signal.
OdaTrak™ System
Key Features
Specifically designed for harsh wastewater environments: unique adapter fully sealed, proven & robust design
Quick & easy installation: standard M20 electrical thread, special machined housing for easy mounting of the OdaTrak™ optic cable
Programmed fault detection: OdaTrak detects loss of data from the OdaLog and will output a fault signal on the 4-20mA line alerting user to fault condition
Special fibre optic cable (ordered separately): safe to use in most commonly found hazardous environments. Two models are available, both with stainless steel connectors; 1. standard coating for the fibre optic cable; 2. extra protective sheath added over the cable for harsher wastewater environments.
Easy to transmit data: incorporates a 4-20mA adapter to transmit gas data using a fibre optic cable
Real time data: readily available when connected to a PLC network