Glyoxalbis(2-hydroxyanil), 97%
Glyoxalbis(2-hydroxyanil), 97%
Glyoxalbis(2-hydroxyanil), 97%
Thermo Scientific Chemicals

Glyoxalbis(2-hydroxyanil), 97%

CAS: 1149-16-2 | C14H12N2O2 | 240.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 120160500
Price (USD)
266.65
Special offer
Online exclusive
Ends: 28-Dec-2026
314.00
Save 47.35
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
266.65
Special offer
Online exclusive
Ends: 28-Dec-2026
314.00
Save 47.35
Each
Chemical Identifiers
CAS1149-16-2
IUPAC Name6-{[(1E)-2-[(2-hydroxyphenyl)amino]ethenyl]imino}cyclohexa-2,4-dien-1-one
Molecular FormulaC14H12N2O2
InChI KeyOMJNMRJMNUREOJ-KOBPNRPCSA-N
SMILESOC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Sulfated ash=<0.1 % (800°C)
Appearance (Color)Beige
HPLC>=96.0 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only