Guaiazulene, 99%
Guaiazulene, 99%
Guaiazulene, 99%
Thermo Scientific Chemicals

Guaiazulene, 99%

CAS: 489-84-9 | C15H18 | 198.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 120200100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS489-84-9
IUPAC Name1,4-dimethyl-7-(propan-2-yl)azulene
Molecular FormulaC15H18
InChI KeyFWKQNCXZGNBPFD-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C)C2=CC=C(C)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue
Infrared spectrumConforms
Appearance (Form)Low melting crystalline solid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only