Nunc™ Automated Cell Factory Manipulator
Thermo Scientific™
Nunc™ Automated Cell Factory Manipulator
The Thermo Scientific Nunc Cell Factory Manipulator is a fully programmable manipulator that automates the handling of Nunc Cell FactoryWeitere Informationen
Have Questions?
KatalognummerTemperaturbereich (metrisch)
1202885 bis 40 °C
Katalognummer 120288
Preis (EUR)
-
Temperaturbereich (metrisch):
5 bis 40 °C
The Thermo Scientific Nunc Cell Factory Manipulator is a fully programmable manipulator that automates the handling of Nunc Cell Factory systems. Automate the filling, washing, emptying, trypsinizing, and harvesting steps of your adherent cell workflows easily and reliably.
• Increase cell-culture processing speed for vaccine or cell/gene therapy manufacturing
• Simplify scale-up while keeping proven adherent cell culture practices
• Efficiently manipulate several layers of standard Nunc Cell Factory systems at the same time to produce adherent cells
Scale-up conveniently
The automated Nunc Cell Factory Manipulator can hold twelve 10-layer to four 40-layer standard Nunc Cell Factory systems (twelve 13-layer or four 52-layer Nunc High Density Cell Factory systems). It comes with a total of 29 programs; 11 fixed basic programs and 18 preinstalled modifiable wash programs.
Consumables customization
Nunc Cell Factory systems can be configured with tubing sets and manifolds to enable a closed system workflow that works seamlessly with the Nunc Automated Cell Factory Manipulator. These tubing sets are customized to your specific needs through our configuration service.
Specifications
Zertifizierung/KonformitätCE Compliance
Elektrische Anforderungen208/400 V, triple phase, 50/60 Hz
Frequenz50/60 Hz
AufnahmevermögenNunc™ Cell Factory™ systems
Modell120288
TypManipulator
Temperaturbereich (metrisch)5 bis 40 °C
Spannung208/400 V
Unit SizeEach