Nalgene™ PMP Griffin Low-Form Plastic Beakers
Nalgene™ PMP Griffin Low-Form Plastic Beakers
Nalgene™ PMP Griffin Low-Form Plastic Beakers
Nalgene™ PMP Griffin Low-Form Plastic Beakers
Thermo Scientific™

Nalgene™ PMP Griffin Low-Form Plastic Beakers

Continuously use these transparent, PMP beakers with easy-to-read, silk-screened graduation at temperatures of 150°C with intermittent service to 175°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Unit Size
1203-4000PK
also known as 1203-4000
4000 mLEach
1203-0030PK
also known as 1203-0030
30 mLPack of 12
1203-005050 mLCase of 36
1203-0050PK
also known as 1203-0050
50 mLPack of 12
1203-0100100 mLCase of 36
1203-0100PK
also known as 1203-0100
100 mLPack of 12
1203-0150150 mLCase of 36
1203-0150PK
also known as 1203-0150
150 mLPack of 12
1203-0250250 mLCase of 24
1203-0250PK
also known as 1203-0250
250 mLPack of 6
1203-0400400 mLCase of 24
1203-0400PK
also known as 1203-0400
400 mLPack of 6
1203-0600600 mLCase of 12
1203-0600PK
also known as 1203-0600
600 mLPack of 4
1203-10001000 mLCase of 12
1203-1000PK
also known as 1203-1000
1000 mLPack of 3
1203-20002000 mLCase of 4
1203-2000PK
also known as 1203-2000
2000 mLEach
1203-40004000 mLCase of 4
Catalog number 1203-4000PK
also known as 1203-4000
Price (USD)
114.07
Online exclusive
126.79
Save 12.72
Each
Capacity (Metric):
4000 mL
Unit Size:
Each
Price (USD)
114.07
Online exclusive
126.79
Save 12.72
Each

Continuously use Thermo Scientific™ Nalgene™ PMP Griffin Low-Form Beakers at temperatures of 150°C with intermittent service to 175°C. These polymethylpentene beakers have the transparency of glass, excellent chemical resistance and heat tolerance. All sizes of beakers have easy-to-read, silk-screened graduations.

  • Transparent PMP material allows for a clear view of beaker contents
  • No-drip, single-spout design provides precision pouring control
  • With ribs for easy stacking
  • Silk screened with graduations, size code, resin code, catalog number, max. temperature and “No Flame” symbol for easy identification and safe use
  • Autoclavable/Graduated/Transparent
  • Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Accuracy
.
Specifications
ColorClear
DescriptionBeaker Griffin Low-Form PMP 4000 mL
Graduation Range500 to 4000 mL
Increments250 mL
Quantity1/Pk.; 4/Cs.
TypeBeaker
AutoclavableAutoclavable
Capacity (Metric)4000 mL
Certifications/ComplianceISO/DIN 7056
DisposableReusable
GraduatedYes
MaterialPMP
Scale500mL to 4000ml in 250mL increments
StyleGriffin, Low Form
Unit SizeEach

Frequently asked questions (FAQs)

Several of your labware products are available in polypropylene (PP, PPCO) or polymethylpentene (PMP). Which should I choose?

These two materials were chosen as labware raw materials for their durability, broad chemical compatibility profile, and wide range of use temperature. Polymethylpentene offers a higher level of transparency (comparable to glass) but is slightly more brittle and therefore somewhat more likely than polypropylene to crack if dropped. Chemical or environmental factors in some applications may strongly indicate the use of only one material- please refer to laboratory plastics reference materials or call Thermo Scientific Laboratory Products Essentials technical support for assistance.