Heptaldehyde, 95%, stabilized
Heptaldehyde, 95%, stabilized
Heptaldehyde, 95%, stabilized
Thermo Scientific Chemicals

Heptaldehyde, 95%, stabilized

CAS: 111-71-7 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 120320010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-71-7
IUPAC Nameheptanal
Molecular FormulaC7H14O
InChI KeyFXHGMKSSBGDXIY-UHFFFAOYSA-N
SMILESCCCCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow to light orange
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
Refractive index1.4100 to 1.4220 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only