n-Heptane, 99+%, pure
n-Heptane, 99+%, pure
n-Heptane, 99+%, pure
Thermo Scientific Chemicals

n-Heptane, 99+%, pure

CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
25 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 120340010
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=99.0 %
Refractive index1.3850 to 1.3890 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only