3-Heptanone, 98%
3-Heptanone, 98%
3-Heptanone, 98%
Thermo Scientific Chemicals

3-Heptanone, 98%

CAS: 106-35-4 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 120380010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106-35-4
IUPAC Nameheptan-3-one
Molecular FormulaC7H14O
InChI KeyNGAZZOYFWWSOGK-UHFFFAOYSA-N
SMILESCCCCC(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4075 to 1.4095 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only