n-Hexadecyl mercaptan, 90%, technical
n-Hexadecyl mercaptan, 90%, technical
n-Hexadecyl mercaptan, 90%, technical
Thermo Scientific Chemicals

n-Hexadecyl mercaptan, 90%, technical

CAS: 2917-26-2 | C16H34S | 258.5 g/mol
Have Questions?
Catalog NumberQuantityPackaging
120521000100 mLGlass bottle
Catalog number 120521000
Price (USD)
95.00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
95.00
Each
Chemical Identifiers
CAS2917-26-2
IUPAC Namehexadecane-1-thiol
Molecular FormulaC16H34S
InChI KeyORTRWBYBJVGVQC-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid or waxy solid
Refractive index1.4610 to 1.4640 (20°C, 589 nm)
Appearance (Color)Slightly hazy colorless to white to light yellow
GC>=88 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only