CHO-S-SFM II, with hypoxanthine and thymidine, 500 mL - FAQs
CHO-S-SFM II, with hypoxanthine and thymidine, 500 mL - FAQs
View additional product information for CHO-S-SFM II, with hypoxanthine and thymidine - FAQs (12052114, 12052098)
1 product FAQs found
Which serum-free medium should I select for my CHO cells?
Thermo Fisher Scientific offers multiple serum free media (SFM) and chemically defined (CD) media options for CHO cultures. For adherent cultures, CD CHO (10743-011) and CD OptiCHO medium (12681-011) are robust media; both are protein free and animal origin free (AOF). For adherent cultures, CD CHO-A (097-0182DJ) is a widely used protein free and animal origin free medium. There are other formulations available to suit other research needs; contact Technical Support for further information.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.