3-Hydroxybenzaldehyd, 98.5 %, Thermo Scientific Chemicals
3-Hydroxybenzaldehyd, 98.5 %, Thermo Scientific Chemicals
3-Hydroxybenzaldehyd, 98.5 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxybenzaldehyd, 98.5 %, Thermo Scientific Chemicals

CAS: 100-83-4 | C7H6O2 | 122.12 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 g
500 g
Verpackung:
Kunststoffflasche
Katalognummer 120965000
Preis (EUR)
188,00
Each
Menge:
500 g
Verpackung:
Kunststoffflasche
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
188,00
Each
Chemikalien-Kennzeichnungen
CAS100-83-4
IUPAC Name3-hydroxybenzaldehyde
Molecular FormulaC7H6O2
InChI KeyIAVREABSGIHHMO-UHFFFAOYSA-N
SMILESOC1=CC=CC(C=O)=C1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Form)Crystalline powder
Melting point101°C to 105°C
Appearance (Color)Light yellow to brown
Infrared spectrumConforms
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only