2-Hydroxy-4-methoxybenzophenone, 98%
2-Hydroxy-4-methoxybenzophenone, 98%
2-Hydroxy-4-methoxybenzophenone, 98%
Thermo Scientific Chemicals

2-Hydroxy-4-methoxybenzophenone, 98%

CAS: 131-57-7 | C14H12O3 | 228.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number 121360050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS131-57-7
IUPAC Name2-benzoyl-5-methoxyphenol
Molecular FormulaC14H12O3
InChI KeyDXGLGDHPHMLXJC-UHFFFAOYSA-N
SMILESCOC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point>=62.0°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only