3-Hydroxy-3-methyl-2-butanone, 92%
3-Hydroxy-3-methyl-2-butanone, 92%
3-Hydroxy-3-methyl-2-butanone, 92%
Thermo Scientific Chemicals

3-Hydroxy-3-methyl-2-butanone, 92%

CAS: 115-22-0 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 121440250
Price (USD)
191.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
191.00
Each
Chemical Identifiers
CAS115-22-0
IUPAC Name3-hydroxy-3-methylbutan-2-one
Molecular FormulaC5H10O2
InChI KeyBNDRWEVUODOUDW-UHFFFAOYSA-N
SMILESCC(=O)C(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow to light red
Infrared spectrumConforms
GC>=90 %
Refractive index1.4140 to 1.4170 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only