2-Hydroxy-p-naphthoquinone, 99%
2-Hydroxy-p-naphthoquinone, 99%
2-Hydroxy-p-naphthoquinone, 99%
Thermo Scientific Chemicals

2-Hydroxy-p-naphthoquinone, 99%

CAS: 83-72-7 | C10H6O3 | 174.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 121630250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS83-72-7
IUPAC Name4-hydroxy-1,2-dihydronaphthalene-1,2-dione
Molecular FormulaC10H6O3
InChI KeyWVCHIGAIXREVNS-UHFFFAOYSA-N
SMILESOC1=CC(=O)C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=98.5 %
Appearance (Color)Yellow to orange to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only