4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%
4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%
4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%
4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%
Thermo Scientific Chemicals

4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%

CAS: 345909-35-5 | C10H7NO3 | 189.17 g/mol
製品番号(カタログ番号) 121910250
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS345909-35-5
IUPAC Name4-hydroxyquinoline-2-carboxylic acid
Molecular FormulaC10H7NO3
InChI KeyHCZHHEIFKROPDY-UHFFFAOYSA-N
SMILESOC(=O)C1=NC2=CC=CC=C2C(O)=C1
さらに表示
Appearance (Color)White to yellow to brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration excess NaOH97.5 to 102.5 % (On dry substance)
Water7 to 10 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only