trans-4-Hydroxystilbene, 98%
trans-4-Hydroxystilbene, 98%
trans-4-Hydroxystilbene, 98%
Thermo Scientific Chemicals

trans-4-Hydroxystilbene, 98%

CAS: 6554-98-9 | C14H12O | 196.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 121930050
Price (USD)
71.65
Online exclusive
79.60
Save 7.95
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
71.65
Online exclusive
79.60
Save 7.95
Each
Chemical Identifiers
CAS6554-98-9
IUPAC Name4-[(1E)-2-phenylethenyl]phenol
Molecular FormulaC14H12O
InChI KeyQVLMUEOXQBUPAH-VOTSOKGWSA-N
SMILESOC1=CC=C(C=CC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point185°C to 189°C
HPLC>=96.5 %
Appearance (Color)Yellow to light brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only