Particle-Certified Glass Containers
Actual product may vary
Particle-Certified Glass Containers
Particle-Certified Glass Containers
Particle-Certified Glass Containers
Particle-Certified Glass Containers
Particle-Certified Glass Containers
Thermo Scientific™

Particle-Certified Glass Containers

Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Product TypeColorNo. per Case
122-16ALP500 mLPacker BottleAmber12
140-40CLP40 mLContainerClear90
120-02ALP60 mLPacker BottleAmber24
114-060ALP60 mLBoston Round BottleAmber24
114-125ALP125 mLBoston Round BottleAmber12
120-04ALP125 mLPacker BottleAmber12
114-125CTLP125 mLBoston Round BottleClear12
121-08ALP250 mLPacker BottleAmber12
114-250ALP250 mLBoston Round BottleAmber12
114-250CLP250 mLBoston Round BottleClear24
131-08CLP
also known as 131-08C/LP
250 mLSample JarClear24
132-16CLP500 mLSample JarClear12
113-500ALP500 mLBoston Round BottleAmber12
112-01ALP1000 mLSample BottleAmber12
133-32CLP1000 mLSample JarClear12
110-80ALP2500 mLContainerAmber6
123-80ALP2500 mLPacker BottleAmber4
111-04ALP4000 mLJugAmber4
Catalog number 122-16ALP
Price (USD)
485.00
Each
Capacity (Metric):
500 mL
Product Type:
Packer Bottle
Color:
Amber
No. per Case:
12
Price (USD)
485.00
Each
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.

Key Features:

  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
  • Individual bottles double bagged to maintain integrity until point of use
  • 4 L jugs suitable for hazardous shipping in conjunction with combination packaging

Recommended for:

  • Low particle applications
Specifications
Capacity16 oz., 500 mL
Capacity (English)16 oz.
Closure LinerPTFE
Closure MaterialPolypropylene, Low-shedding
Closure Size53-400 mm
DescriptionAmber Glass Wide Mouth Packer Bottle with Closure, 500 mL
Diameter (English)5.8 in.
Diameter (Metric)147.32 mm
Height (English)3.18 in.
Height (Metric)80.77 mm
No. per Case12
Product TypePacker Bottle
Capacity (Metric)500 mL
ColorAmber
MaterialGlass
MouthWide
Unit SizeEach